5-Cyclopropylpyridine-2-carbonitrile

Supplier: Apollo Scientific

OR315867-1G OR315867-250MG
APOSOR315867-1GEA 493 EUR
APOSOR315867-1G APOSOR315867-250MG
5-Cyclopropylpyridine-2-carbonitrile
5-Cyclopropylpyridine-2-carbonitrile
Formula: C₉H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD13689136
CAS Number: 188918-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR