2-(Piperidin-3-yl)ethan-1-ol hydrochloride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(3-Piperidyl)ethanol Hydrochloride
OR321304-1G OR321304-5G OR321304-10G
APOSOR321304-1GEA 53.8 EUR
APOSOR321304-1G APOSOR321304-5G APOSOR321304-10G
2-(Piperidin-3-yl)ethan-1-ol hydrochloride 97%
2-(Piperidin-3-yl)ethan-1-ol hydrochloride
Formula: C₇H₁₅NO·ClH
MW: 165.661 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14583117
CAS Number: 16780-04-4