3,6-Dimethylpyrazin-2-amine 97%
Supplier: Apollo Scientific
Warning
OR321371-1G OR321371-250MG
APOSOR321371-1GEA 69.6 EUR
APOSOR321371-1G APOSOR321371-250MG
3,6-Dimethylpyrazin-2-amine 97%
3,6-Dimethylpyrazin-2-amine
Formula: C₆H₉N₃
MW: 123.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917551
CAS Number: 13134-38-8