4-(Piperidin-4-yl)benzonitrile hydrochloride
Supplier: Apollo Scientific
OR321508-1G OR321508-250MG
APOSOR321508-1GEA 589 EUR
APOSOR321508-1G APOSOR321508-250MG
4-(Piperidin-4-yl)benzonitrile hydrochloride
4-(Piperidin-4-yl)benzonitrile hydrochloride
Formula: C₁₂H₁₄N₂·ClH
MW: 222.714 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08544124
CAS Number: 162997-34-4