7-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine, N1-BOC protected 95%
Supplier: Apollo Scientific
Warning
OR33671-1G OR33671-250MG
APOSOR33671-1GEA 783 EUR
APOSOR33671-1G APOSOR33671-250MG
7-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine, N1-BOC protected 95%
7-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine, N1-BOC protected
Formula: C₁₃H₁₇BrN₂O₂
MW: 313.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21605206
CAS Number: 1375301-99-7