Methyl 2-bromo-3-methoxypropanoate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-bromo-3-methoxypropionate
OR3386-25G OR3386-5G
APOSOR3386-25GEA 468 EUR
APOSOR3386-25G APOSOR3386-5G
Methyl 2-bromo-3-methoxypropanoate 95%
Methyl 2-bromo-3-methoxypropanoate
Formula: C₅H₉BrO₃
MW: 197.027 g/mol
Boiling Pt: 69 °C (11 mm Hg)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD07784248
CAS Number: 27704-96-7
UN: 1993
ADR: 3 III