2,6-Dibromo-4-methoxypyridine 99%
Supplier: Apollo Scientific
Warning
OR345215-1G OR345215-5G
APOSOR345215-1GEA 81.2 EUR
APOSOR345215-1G APOSOR345215-5G
2,6-Dibromo-4-methoxypyridine 99%
2,6-Dibromo-4-methoxypyridine
Formula: C₆H₅Br₂NO
Melting Pt: 137…139 °C
Storage Temperature: Ambient
MDL Number: MFCD11036227
CAS Number: 117873-72-0