3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
OR346226-1G OR346226-5G
APOSOR346226-1GEA 0 EUR
APOSOR346226-1G APOSOR346226-5G
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid
3-Methyl-5-p-tolyl-isoxazole-4-carboxylic acid
Formula: C₁₂H₁₁NO₃
MW: 217.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11557830
CAS Number: 91569-57-2