2,2-Dimethyl-5-phenyl-[1,3]dioxolan-4-one
Supplier: Apollo Scientific
OR346236-1G OR346236-500MG OR346236-5G
APOSOR346236-1GEA 0 EUR
APOSOR346236-1G APOSOR346236-500MG APOSOR346236-5G
2,2-Dimethyl-5-phenyl-[1,3]dioxolan-4-one
2,2-Dimethyl-5-phenyl-[1,3]dioxolan-4-one
Formula: C₁₁H₁₂O₃
MW: 192.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01327620
CAS Number: 6337-34-4