1-tert-Butoxycarbonyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)pyrazole
Supplier: Apollo Scientific
Synonyms: tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate
OR361611-250MG OR361611-1G
APOSOR361611-250MGEA 151 EUR
APOSOR361611-250MG APOSOR361611-1G
1-tert-Butoxycarbonyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)pyrazole
1-tert-Butoxycarbonyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)pyrazole
Formula: C₁₄H₂₃BN₂O₄
MW: 294.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368045
CAS Number: 914672-66-5