4-(Bromomethyl)-3,5-dibromobenzoic acid 95%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Carboxy-2,6-dibromobenzyl bromide, α,3,5-Tribromo-p-toluic acid
OR400310-1G OR400310-250MG
APOSOR400310-1GEA 348 EUR
APOSOR400310-1G APOSOR400310-250MG
4-(Bromomethyl)-3,5-dibromobenzoic acid 95%
4-(Bromomethyl)-3,5-dibromobenzoic acid
Formula: C₈H₅Br₃O₂
MW: 372.836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28096537
CAS Number: 1820607-63-3
UN: 3261
ADR: 8 III