2-Amino-4-bromo-6-chlorophenol
Supplier: Apollo Scientific
OR400332-1G OR400332-250MG OR400332-5G
APOSOR400332-1GEA 77 EUR
APOSOR400332-1G APOSOR400332-250MG APOSOR400332-5G
2-Amino-4-bromo-6-chlorophenol
2-Amino-4-bromo-6-chlorophenol
Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16742724
CAS Number: 855836-14-5