5-Methylpyrazine-2-carboxaldehyde 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Methylpyrazine-2-carbaldehyde, 2-Formyl-5-methylpyrazine, 2-Formyl-5-methyl-1,4-diazine
OR40083-1G OR40083-5G OR40083-25G
APOSOR40083-1GEA 53.8 EUR
APOSOR40083-1G APOSOR40083-5G APOSOR40083-25G
5-Methylpyrazine-2-carboxaldehyde 95%
5-Methylpyrazine-2-carboxaldehyde
Formula: C₆H₆N₂O
MW: 122.125 g/mol
Boiling Pt: 38…40 °C (1 mm Hg)
Melting Pt: 42278 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10697792
CAS Number: 50866-30-3