4-Bromo-3-chlorostyrene 95%
Supplier: Apollo Scientific
OR400918-1G OR400918-250MG
APOSOR400918-1GEA 609 EUR
APOSOR400918-1G APOSOR400918-250MG
4-Bromo-3-chlorostyrene 95%
4-Bromo-3-chlorostyrene
Formula: C₈H₆BrCl
Storage Temperature: Refrigerator
MDL Number: MFCD30237713
CAS Number: 1692077-74-9