2'-Bromo-6'-chloroacetophenone 95%

Supplier: Apollo Scientific
Warning

OR400923-1G OR400923-5G OR400923-25G
APOSOR400923-1GEA 72.5 EUR
APOSOR400923-1G APOSOR400923-5G APOSOR400923-25G
2'-Bromo-6'-chloroacetophenone 95%
2'-Bromo-6'-chloroacetophenone
Formula: C₈H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD18390298
CAS Number: 1261438-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR