3-(4-Bromophenyl)-2-methylpropanoic acid
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-Bromobenzyl)propanoic acid, 3-(4-Bromophenyl)-2-methylpropionic acid
OR40318-1G OR40318-250MG OR40318-100MG
APOSOR40318-1GEA 415 EUR
APOSOR40318-1G APOSOR40318-250MG APOSOR40318-100MG
3-(4-Bromophenyl)-2-methylpropanoic acid
3-(4-Bromophenyl)-2-methylpropanoic acid
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12152839
CAS Number: 66735-01-1