3-Chloro-2-(hydroxymethyl)-6-methoxypyridine
Supplier: Apollo Scientific
Warning
Synonyms: (3-Chloro-6-methoxypyridin-2-yl)methanol, 3-Chloro-6-methoxypicolinyl alcohol
OR40364-1G OR40364-250MG
APOSOR40364-1GEA 212 EUR
APOSOR40364-1G APOSOR40364-250MG
3-Chloro-2-(hydroxymethyl)-6-methoxypyridine
3-Chloro-2-(hydroxymethyl)-6-methoxypyridine
Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
Boiling Pt: 100…102 °C (0.3 mm Hg)
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD14606397
CAS Number: 1227490-30-3