2,6-Dimethylanthraquinone 90%
Supplier: Apollo Scientific
Synonyms: 2,6-Dimethylanthracene-9,10-dione
OR42110-1G OR42110-250MG
APOSOR42110-1GEA 1450 EUR
APOSOR42110-1G APOSOR42110-250MG
2,6-Dimethylanthraquinone 90%
2,6-Dimethylanthraquinone
Formula: C₁₆H₁₂O₂
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00835218
CAS Number: 3837-38-5