3-Bromo-6-nitroquinoline
Supplier: Apollo Scientific
Synonyms: 3-Bromo-6-nitro-1-azanaphthalene
OR45087-1G OR45087-250MG
APOSOR45087-1GEA 575 EUR
APOSOR45087-1G APOSOR45087-250MG
3-Bromo-6-nitroquinoline
3-Bromo-6-nitroquinoline
Formula: C₉H₅BrN₂O₂
Melting Pt: 165…171 °C
Storage Temperature: Ambient
MDL Number: MFCD12024480
CAS Number: 7101-95-3