2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
Supplier: Apollo Scientific
Danger
OR451401-1G OR451401-5G
APOSOR451401-1GEA 102 EUR
APOSOR451401-1G APOSOR451401-5G
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine dihydrochloride
Formula: C₉H₁₀ClN₃·2ClH
MW: 268.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419439
CAS Number: 91003-86-0
UN: 3259
ADR: 8 III