4-Chloro-1-methyl-1H-pyrazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
OR451455-1G OR451455-25G OR451455-5G
APOSOR451455-1GEA 139 EUR
APOSOR451455-1G APOSOR451455-25G APOSOR451455-5G
4-Chloro-1-methyl-1H-pyrazole-5-carboxylic acid
4-Chloro-1-methyl-1H-pyrazole-5-carboxylic acid
Formula: C₅H₅ClN₂O₂
MW: 160.558 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463989
CAS Number: 84547-83-1