3-Morpholineethanol hydrochloride 97%
Supplier: Apollo Scientific
OR452075-100MG OR452075-250MG OR452075-500MG
APOSOR452075-100MGEA 328 EUR
APOSOR452075-100MG APOSOR452075-250MG APOSOR452075-500MG
3-Morpholineethanol hydrochloride 97%
3-Morpholineethanol hydrochloride
Formula: C₆H₁₃NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD22199183
CAS Number: 399580-64-4