N-Boc-(R)-2-(methylamino)butyric acid 97%
Supplier: Apollo Scientific
OR452090-1G OR452090-500MG
APOSOR452090-1GEA 393 EUR
APOSOR452090-1G APOSOR452090-500MG
N-Boc-(R)-2-(methylamino)butyric acid 97%
N-Boc-(R)-2-(methylamino)butyric acid
Formula: C₁₀H₁₉NO₄
MW: 217.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380682
CAS Number: 287210-80-4