N-Methylisonipecotic acid, ethyl ester, hydrochloride
Supplier: Apollo Scientific
Danger
OR460036-1G OR460036-25G OR460036-5G
APOSOR460036-1GEA 90.5 EUR
APOSOR460036-1G APOSOR460036-25G APOSOR460036-5G
N-Methylisonipecotic acid, ethyl ester, hydrochloride
N-Methylisonipecotic acid, ethyl ester, hydrochloride
Formula: C₉H₁₇NO₂·ClH
MW: 207.698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12965068
CAS Number: 52700-61-5