2-Isopropoxy-1,3-thiazole-5-boronic acid, pinacol ester
Supplier: Apollo Scientific
Warning
Synonyms: 2-[(Prop-2-yl)oxy]-1,3-thiazole-5-boronic acid, pinacol ester, 2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole, 2-[(Prop-2-yl)oxy]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
OR46023-1G OR46023-250MG
APOSOR46023-1GEA 799 EUR
APOSOR46023-1G APOSOR46023-250MG
2-Isopropoxy-1,3-thiazole-5-boronic acid, pinacol ester
2-Isopropoxy-1,3-thiazole-5-boronic acid, pinacol ester
Formula: C₁₂H₂₀BNO₃S
MW: 269.168 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032347
CAS Number: 1352652-22-2