2-Methyl-1-phenyl-1H-benzimidazole-5-carboxylic acid
Supplier: Apollo Scientific
Warning
Synonyms: 5-Carboxy-2-methyl-1-phenyl-1H-benzimidazole, 2-Methyl-1-phenyl-1H-benzo[d]imidazole-5-carboxylic acid
OR46085-1G OR46085-5G
APOSOR46085-1GEA 270 EUR
APOSOR46085-1G APOSOR46085-5G
2-Methyl-1-phenyl-1H-benzimidazole-5-carboxylic acid
2-Methyl-1-phenyl-1H-benzimidazole-5-carboxylic acid
Formula: C₁₅H₁₂N₂O₂
MW: 252.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00218479
CAS Number: 92437-43-9