3-Methylenecyclobutanamine hydrochloride

Supplier: Apollo Scientific
Warning

OR46630-1G OR46630-100MG OR46630-250MG OR46630-500MG OR46630-2.5G OR46630-5G
APOSOR46630-1GEA 470 EUR
APOSOR46630-1G APOSOR46630-100MG APOSOR46630-250MG APOSOR46630-500MG APOSOR46630-2.5G APOSOR46630-5G
3-Methylenecyclobutanamine hydrochloride
3-Methylenecyclobutanamine hydrochloride
Formula: C₅H₉N·ClH
Storage Temperature: Ambient
MDL Number: MFCD26404481
CAS Number: 136137-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR