3-(3-Nitrophenoxy)propylamine

Supplier: Apollo Scientific

Synonyms: 3-(3-Nitrophenoxy)propan-1-amine

OR470096-1G OR470096-5G
APOSOR470096-1GEA 361 EUR
APOSOR470096-1G APOSOR470096-5G
3-(3-Nitrophenoxy)propylamine
3-(3-Nitrophenoxy)propylamine
Formula: C₉H₁₂N₂O₃
MW: 196.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12818946
CAS Number: 116753-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR