(R)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine
Supplier: Apollo Scientific
OR470121-1G OR470121-5G
APOSOR470121-1GEA 275 EUR
APOSOR470121-1G APOSOR470121-5G
(R)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine
(R)-N-Methyl-N-(2-propynyl)-2,3-dihydroinden-1-amine
Formula: C₁₃H₁₅N
MW: 185.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367857
CAS Number: 124192-87-6