Pentanimidamide

Supplier: Apollo Scientific

OR470546-1G OR470546-5G
APOSOR470546-1GEA 357 EUR
APOSOR470546-1G APOSOR470546-5G
Pentanimidamide
Pentanimidamide
Formula: C₅H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD05663348
CAS Number: 109-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR