1,2-Bis(4-chlorophenyl)ethylenediamine
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Bis(4-chlorophenyl)ethane-1,2-diamine
OR471166-5G OR471166-1G
APOSOR471166-5GEA 916 EUR
APOSOR471166-5G APOSOR471166-1G
1,2-Bis(4-chlorophenyl)ethylenediamine
1,2-Bis(4-chlorophenyl)ethylenediamine
Formula: C₁₄H₁₄Cl₂N₂
MW: 281.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05150371
CAS Number: 86212-34-2