7-Chloro-1-methyl-1,8-naphthyridin-2(1H)-one
Supplier: Apollo Scientific
OR471315-1G OR471315-250MG
APOSOR471315-1GEA 1120 EUR
APOSOR471315-1G APOSOR471315-250MG
7-Chloro-1-methyl-1,8-naphthyridin-2(1H)-one
7-Chloro-1-methyl-1,8-naphthyridin-2(1H)-one
Formula: C₉H₇ClN₂O
MW: 194.618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28501517
CAS Number: 69099-05-4