4-(Hydroxymethyl)oxazolidin-2-one
Supplier: Apollo Scientific
OR471489-1G OR471489-5G
APOSOR471489-1GEA 158 EUR
APOSOR471489-1G APOSOR471489-5G
4-(Hydroxymethyl)oxazolidin-2-one
4-(Hydroxymethyl)oxazolidin-2-one
Formula: C₄H₇NO₃
MW: 117.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045779
CAS Number: 15546-08-4