3,6-Dibromo-4-methylpyridazine
Supplier: Apollo Scientific
OR471593-1G OR471593-5G
APOSOR471593-1GEA 58 EUR
APOSOR471593-1G APOSOR471593-5G
3,6-Dibromo-4-methylpyridazine
3,6-Dibromo-4-methylpyridazine
Formula: C₅H₄Br₂N₂
MW: 251.907 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056210
CAS Number: 89284-10-6