5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 95%
Supplier: Apollo Scientific
Warning
OR471704-1G OR471704-250MG
APOSOR471704-1GEA 51.5 EUR
APOSOR471704-1G APOSOR471704-250MG
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine 95%
5-Boc-3-Iodo-4,5,6,7-tetrahydropyrazolo[4,3-c]pyridine
Formula: C₁₁H₁₆IN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27978466
CAS Number: 661487-17-8