(E)-3-Butoxy-1-(3-chlorophenyl)prop-2-en-1-one
Supplier: Apollo Scientific
OR480286-1G OR480286-250MG OR480286-500MG
APOSOR480286-1GEA 605 EUR
APOSOR480286-1G APOSOR480286-250MG APOSOR480286-500MG
(E)-3-Butoxy-1-(3-chlorophenyl)prop-2-en-1-one
(E)-3-Butoxy-1-(3-chlorophenyl)prop-2-en-1-one
Formula: C₁₃H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD22683244
CAS Number: 1375008-14-2