N,N'-(Benzene-1,4-diyl)dibenzenesulphonamide
Supplier: Apollo Scientific
Warning
Synonyms: N1,N4-Bis(phenylsulphonyl)benzene-1,4-diamine, N,N'-(1,4-Phenylene)dibenzenesulphonamide
OR48246-1G OR48246-250MG OR48246-50MG
APOSOR48246-1GEA 418 EUR
APOSOR48246-1G APOSOR48246-250MG APOSOR48246-50MG
N,N'-(Benzene-1,4-diyl)dibenzenesulphonamide
N,N'-(Benzene-1,4-diyl)dibenzenesulphonamide
Formula: C₁₈H₁₆N₂O₄S₂
MW: 388.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00183146
CAS Number: 16504-19-1