4-(Azidoacetyl)morpholine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Azido-1-(morpholin-4-yl)ethan-1-one, 2-(Morpholin-4-yl)-2-oxoethyl azide
OR4970-1G OR4970-5G OR4970-500MG OR4970-250MG
APOSOR4970-1GEA 364 EUR
APOSOR4970-1G APOSOR4970-5G APOSOR4970-500MG APOSOR4970-250MG
4-(Azidoacetyl)morpholine
4-(Azidoacetyl)morpholine
Formula: C₆H₁₀N₄O₂
MW: 170.169 g/mol
Boiling Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD01320911
CAS Number: 864528-04-1