3-Methyl-1-[(methylsulphanyl)methanethioyl]-1H-imidazol-3-ium iodide
Supplier: Apollo Scientific
OR50518-1G OR50518-250MG OR50518-5G
APOSOR50518-1GEA 232 EUR
APOSOR50518-1G APOSOR50518-250MG APOSOR50518-5G
3-Methyl-1-[(methylsulphanyl)methanethioyl]-1H-imidazol-3-ium iodide
3-Methyl-1-[(methylsulphanyl)methanethioyl]-1H-imidazol-3-ium iodide
Formula: C₆H₉N₂S₂·I
MW: 300.184 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD32648175
CAS Number: 262350-35-6