tert-Butyl 3-aminopropanoate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 3-Aminopropanoate
OR51868-1G OR51868-250MG OR51868-5G
APOSOR51868-1GEA 105 EUR
APOSOR51868-1G APOSOR51868-250MG APOSOR51868-5G
tert-Butyl 3-aminopropanoate
tert-Butyl 3-aminopropanoate
Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Boiling Pt: 192.3 °C
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD06212562
CAS Number: 15231-41-1