4-Acetoxybenzyl alcohol
Supplier: Apollo Scientific
Warning
OR51991-1G OR51991-5G OR51991-10G
APOSOR51991-1GEA 75.4 EUR
APOSOR51991-1G APOSOR51991-5G APOSOR51991-10G
4-Acetoxybenzyl alcohol
4-Acetoxybenzyl alcohol
Formula: C₉H₁₀O₃
Melting Pt: 37…42 °C
Density: 1.169 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02752162
CAS Number: 6309-46-2