4-Bromophenylhydrazinium chloride 95%
Supplier: Apollo Scientific
Danger
Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride
OR5273-100G OR5273-500G
APOSOR5273-100GEA 77 EUR
APOSOR5273-100G APOSOR5273-500G
4-Bromophenylhydrazinium chloride 95%
4-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Melting Pt: 211…214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012941
CAS Number: 622-88-8
UN: 3261
ADR: 8 II