N-Phenylaminosulphonamide
Supplier: Apollo Scientific
OR54584-1G OR54584-250MG
APOSOR54584-1GEA 605 EUR
APOSOR54584-1G APOSOR54584-250MG
N-Phenylaminosulphonamide
N-Phenylaminosulphonamide
Formula: C₆H₈N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD20483446
CAS Number: 15959-53-2