5-Amino-2,3-dimethoxypyridine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 5,6-Dimethoxypyridin-3-amine
OR55347-100MG OR55347-250MG
APOSOR55347-100MGEA 510 EUR
APOSOR55347-100MG APOSOR55347-250MG
5-Amino-2,3-dimethoxypyridine 95%
5-Amino-2,3-dimethoxypyridine
Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20715092
CAS Number: 79491-49-9