4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,5,5-Tetramethyl-2-[4-(oxiran-2-ylmethoxy)phenyl]-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(4-(oxiran-2-ylmethoxy)phenyl)-1,3,2-dioxaborolane
OR5962-1G OR5962-5G OR5962-10G
APOSOR5962-1GEA 140 EUR
APOSOR5962-1G APOSOR5962-5G APOSOR5962-10G
4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester 98%
4-(Oxiran-2-ylmethoxy)benzeneboronic acid, pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD09027286
CAS Number: 664991-83-7