(11-Bromoundec-1-yl)(trimethoxy)silane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 11-(Trimethoxysilyl)undecyl bromide, (11-Bromoundecyl)trimethoxysilane, 1-Bromo-11-(trimethoxysilyl)undecane
OR61604-10G OR61604-25G OR61604-1G OR61604-5G
APOSOR61604-10GEA 502 EUR
APOSOR61604-10G APOSOR61604-25G APOSOR61604-1G APOSOR61604-5G
(11-Bromoundec-1-yl)(trimethoxy)silane 97%
(11-Bromoundec-1-yl)(trimethoxy)silane
Formula: C₁₄H₃₁BrO₃Si
MW: 355.384 g/mol
Boiling Pt: 158 °C (0.8 mm Hg)
Density: 1.12 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00270978
CAS Number: 17947-99-8