4-[5-(Hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine, N1-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine
OR6759-1G OR6759-250MG
APOSOR6759-1GEA 154 EUR
APOSOR6759-1G APOSOR6759-250MG
4-[5-(Hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine, N1-BOC protected
4-[5-(Hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine, N1-BOC protected
Formula: C₁₅H₂₄N₂O₃S
MW: 312.428 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD07368658
CAS Number: 857283-66-0