1-Methyl-5-[(methylamino)methyl]-1H-imidazole
Supplier: Apollo Scientific
Danger
Synonyms: N-Methyl-N-[(1-methyl-1H-imidazol-5-yl)methyl]amine
OR6804-1G OR6804-250MG
APOSOR6804-1GEA 751 EUR
APOSOR6804-1G APOSOR6804-250MG
1-Methyl-5-[(methylamino)methyl]-1H-imidazole
1-Methyl-5-[(methylamino)methyl]-1H-imidazole
Formula: C₆H₁₁N₃
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD07772801
CAS Number: 384821-19-6
UN: 1760
ADR: 8,III