4-[3-(Piperazin-1-yl)prop-1-yl]morpholine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[3-(Morpholin-4-yl)prop-1-yl]piperazine
OR6915-2G OR6915-5G
APOSOR6915-2GEA 125 EUR
APOSOR6915-2G APOSOR6915-5G
4-[3-(Piperazin-1-yl)prop-1-yl]morpholine 98%
4-[3-(Piperazin-1-yl)prop-1-yl]morpholine
Formula: C₁₁H₂₃N₃O
MW: 213.32 g/mol
Boiling Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01075238
CAS Number: 436852-18-5