Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate
Supplier: Apollo Scientific
Warning
Synonyms: 3-[4-(Methoxycarbonyl)phenyl]prop-2-yn-1-ol
OR7570-250MG OR7570-1G
APOSOR7570-250MGEA 31.9 EUR
APOSOR7570-250MG APOSOR7570-1G
Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate
Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate
Formula: C₁₁H₁₀O₃
MW: 190.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00168848
CAS Number: 61266-36-2